trans-4'-Bromochalcone

Lieferant: Apollo Scientific
Achtung

Synonyme: (2E)-1-(4-Bromophenyl)-3-phenylprop-2-en-1-one

OR27786-1G OR27786-250MG
APOSOR27786-1GEA 143 EUR
APOSOR27786-1G APOSOR27786-250MG
trans-4'-Bromochalcone
trans-4'-Bromochalcone
Formel: C₁₅H₁₁BrO
Molecular Weight: 287.151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00017841
CAS-Nummer: 22966-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR