(2,3-Dihydro-3-oxo-4H-1,4-benzothiazin-4-yl)acetic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(2,3-Dihydro-3-oxo-4H-1,4-benzothiazin-4-yl)ethanoic acid, 4-(Carboxymethyl)-3,4-dihydro-3-oxo-2H-1,4-benzothiazine
OR2786-1G
APOSOR2786-1GEA
161
EUR
APOSOR2786-1G
(2,3-Dihydro-3-oxo-4H-1,4-benzothiazin-4-yl)acetic acid
(2,3-Dihydro-3-oxo-4H-1,4-benzothiazin-4-yl)acetic acid
Formel:
C₁₀H₉NO₃S Molecular Weight: 223,248 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00665896 CAS-Nummer: 100637-60-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...