2,5-Dimethyl-1H-benzimidazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,5-Dimethyl-1H-benzo[d]imidazole

OR28395-10G OR28395-50G OR28395-5G
APOSOR28395-10GEA 290 EUR
APOSOR28395-10G APOSOR28395-50G APOSOR28395-5G
2,5-Dimethyl-1H-benzimidazole
2,5-Dimethyl-1H-benzimidazole
Formel: C₉H₁₀N₂
Molecular Weight: 146,189 g/mol
Schmelzpunkt: 202…205 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00125906
CAS-Nummer: 1792-41-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR