2-[2-(Dimethylamino)ethyl]pyridine

Lieferant: Apollo Scientific
Gefahr

Synonyme: N,N-Dimethyl-2-(pyridin-2-yl)ethylamine, N-[2-(Pyridin-2-yl)ethyl]dimethylamine

OR28892-1G OR28892-5G
APOSOR28892-1GEA 31.9 EUR
APOSOR28892-1G APOSOR28892-5G
2-[2-(Dimethylamino)ethyl]pyridine
2-[2-(Dimethylamino)ethyl]pyridine
Formel: C₉H₁₄N₂
Molecular Weight: 150,221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00023546
CAS-Nummer: 6304-27-4
EINECS: 228-608-3
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR