N-(4-amino-5-methoxy-2-methylphenyl)benzamide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(4-Amino-5-methoxy-2-methylphenyl)benzamide
OR28961-1G
OR28961-250MG
OR28961-500MG
APOSOR28961-1GEA
314
EUR
APOSOR28961-1G
APOSOR28961-250MG
APOSOR28961-500MG
N-(4-amino-5-methoxy-2-methylphenyl)benzamide
N-(4-amino-5-methoxy-2-methylphenyl)benzamide
Formel:
C₁₅H₁₆N₂O₂ Molecular Weight: 256,3 g/mol Schmelzpunkt: 185…188 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064435 CAS-Nummer: 99-21-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...