5-tert-Butyl-3-(chloromethyl)-1,2,4-oxadiazole

Lieferant: Apollo Scientific
Gefahr

OR28995-1G OR28995-250MG
APOSOR28995-1GEA 98.6 EUR
APOSOR28995-1G APOSOR28995-250MG
5-tert-Butyl-3-(chloromethyl)-1,2,4-oxadiazole
5-tert-Butyl-3-(chloromethyl)-1,2,4-oxadiazole
Formel: C₇H₁₁ClN₂O
Molecular Weight: 174,628 g/mol
Siedepunkt: 95…100 °C (10 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00068196
CAS-Nummer: 175205-41-1
UN: 1760
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR