4-(1H-Pyrazol-3-yl)benzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR29-100MG
OR29-1G
APOSOR29-100MGEA
30.6
EUR
APOSOR29-100MG
APOSOR29-1G
4-(1H-Pyrazol-3-yl)benzoic acid
4-(1H-Pyrazol-3-yl)benzoic acid
Formel:
C₁₀H₈N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01569442 CAS-Nummer: 208511-67-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...