Ethyl 5-phenyl-1,3-oxazole-4-carboxylate

Lieferant: Apollo Scientific

Synonyme: [4-(Ethoxycarbonyl)-1,3-oxazol-5-yl]benzene, 4-(Ethoxycarbonyl)-5-phenyl-1,3-oxazole

OR29060-1G OR29060-250MG
APOSOR29060-1GEA 168 EUR
APOSOR29060-1G APOSOR29060-250MG
Ethyl 5-phenyl-1,3-oxazole-4-carboxylate
Ethyl 5-phenyl-1,3-oxazole-4-carboxylate
Formel: C₁₂H₁₁NO₃
Molecular Weight: 217,221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00085084
CAS-Nummer: 32998-97-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR