5-Hydroxy-3-nitrobenzo[b]furan 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Nitro-1-benzofuran-5-ol, 3-Nitrobenzo[b]furan-5-ol, 5-Hydroxy-3-nitro-1-benzofuran, 3-Nitrobenzofuran-5-ol

OR29193-1G OR29193-500MG
APOSOR29193-1GEA 92.8 EUR
APOSOR29193-1G APOSOR29193-500MG
5-Hydroxy-3-nitrobenzo[b]furan 97%
5-Hydroxy-3-nitrobenzo[b]furan
Formel: C₈H₅NO₄
Molecular Weight: 179.13 g/mol
Schmelzpunkt: 164…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00114881
CAS-Nummer: 126318-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR