3-(2,5-Dimethylfur-3-yl)-3-oxopropanenitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Cyanoacetyl)-2,5-dimethylfuran, 3-(2,5-Dimethylfur-3-yl)-3-oxopropionitrile

OR29470-250MG OR29470-50MG
APOSOR29470-250MGEA 279 EUR
APOSOR29470-250MG APOSOR29470-50MG
3-(2,5-Dimethylfur-3-yl)-3-oxopropanenitrile
3-(2,5-Dimethylfur-3-yl)-3-oxopropanenitrile
Formel: C₉H₉NO₂
Molecular Weight: 163,173 g/mol
Schmelzpunkt: 120…122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00275552
CAS-Nummer: 175276-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR