2-(2,3-Dihydrobenzo[b]furan-5-yl)-1,3-thiazole-4-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Carboxy-2-(2,3-dihydro-1-benzofuran-5-yl)-1,3-thiazole, 2-(2,3-Dihydro-1-benzofuran-5-yl)-1,3-thiazole-4-carboxylic acid

OR29479-1G OR29479-5G OR29479-250MG
APOSOR29479-1GEA 185 EUR
APOSOR29479-1G APOSOR29479-5G APOSOR29479-250MG
2-(2,3-Dihydrobenzo[b]furan-5-yl)-1,3-thiazole-4-carboxylic acid 97%
2-(2,3-Dihydrobenzo[b]furan-5-yl)-1,3-thiazole-4-carboxylic acid
Formel: C₁₂H₉NO₃S
Schmelzpunkt: 210…214 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01313485
CAS-Nummer: 368869-97-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR