5-(1-Chloro-2-methylprop-2-yl)-3-(3-methylphenyl)-1,2,4-oxadiazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-[5-(1-Chloro-2-methylprop-2-yl)-1,2,4-oxadiazol-3-yl]toluene, 1-[5-(1-Chloro-2-methylprop-2-yl)-1,2,4-oxadiazol-3-yl]-3-methylbenzene

OR29506-1G OR29506-250MG
APOSOR29506-1GEA 434 EUR
APOSOR29506-1G APOSOR29506-250MG
5-(1-Chloro-2-methylprop-2-yl)-3-(3-methylphenyl)-1,2,4-oxadiazole
5-(1-Chloro-2-methylprop-2-yl)-3-(3-methylphenyl)-1,2,4-oxadiazole
Formel: C₁₃H₁₅ClN₂O
Molecular Weight: 250.724 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01765337
CAS-Nummer: 680216-08-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR