1-(3,5-Dichlorophenyl)-5-propyl-1H-pyrazole-4-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR29530-1G
OR29530-250MG
APOSOR29530-1GEA
0
EUR
APOSOR29530-1G
APOSOR29530-250MG
1-(3,5-Dichlorophenyl)-5-propyl-1H-pyrazole-4-carboxylic acid
1-(3,5-Dichlorophenyl)-5-propyl-1H-pyrazole-4-carboxylic acid
Formel:
C₁₃H₁₂Cl₂N₂O₂ Schmelzpunkt: 169…171 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02677727 CAS-Nummer: 306936-60-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...