Ethyl 6-(chloromethyl)-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylate

Lieferant: Apollo Scientific
Gefahr

Synonyme: 6-(Chloromethyl)-5-(ethoxycarbonyl)-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine, [6-(Chloromethyl)-5-(ethoxycarbonyl)-2-oxo-1,2,3,4-tetrahydropyrimidin-4-yl]benzene

OR29641-1G OR29641-250MG OR29641-500MG
APOSOR29641-1GEA 574 EUR
APOSOR29641-1G APOSOR29641-250MG APOSOR29641-500MG
Ethyl 6-(chloromethyl)-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylate
Ethyl 6-(chloromethyl)-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylate
Formel: C₁₄H₁₅ClN₂O₃
Molecular Weight: 294,734 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00103423
CAS-Nummer: 263157-08-0
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR