Ethyl 4-chloro-2-(thien-2-yl)pyrimidine-5-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-[4-Chloro-5-(ethoxycarbonyl)pyrimidin-2-yl]thiophene, 4-Chloro-5-(ethoxycarbonyl)-2-(thien-2-yl)pyrimidine
OR29803-1G
OR29803-500MG
APOSOR29803-1GEA
551
EUR
APOSOR29803-1G
APOSOR29803-500MG
Ethyl 4-chloro-2-(thien-2-yl)pyrimidine-5-carboxylate
Ethyl 4-chloro-2-(thien-2-yl)pyrimidine-5-carboxylate
Formel:
C₁₁H₉ClN₂O₂S Molecular Weight: 268.719 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00662782 CAS-Nummer: 188937-40-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...