1-Chloroisoquinoline-6-boronic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Borono-1-chloro-2-azanaphthalene, (1-Chloroisoquinolin-6-yl)boronic acid, 6-Borono-1-chloroisoquinoline
OR300002-1G
OR300002-250MG
APOSOR300002-1GEA
546
EUR
APOSOR300002-1G
APOSOR300002-250MG
1-Chloroisoquinoline-6-boronic acid
1-Chloroisoquinoline-6-boronic acid
Formel:
C₉H₇BClNO₂ Molecular Weight: 207.421 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD13190242 CAS-Nummer: 1800484-01-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...