1-Chloroisoquinoline-8-boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Borono-1-chloroisoquinoline, 8-Borono-1-chloro-2-azanaphthalene

OR300004-250MG OR300004-500MG
APOSOR300004-250MGEA 125 EUR
APOSOR300004-250MG APOSOR300004-500MG
1-Chloroisoquinoline-8-boronic acid
1-Chloroisoquinoline-8-boronic acid
Formel: C₉H₇BClNO₂
Molecular Weight: 207,421 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18257024
CAS-Nummer: 2096337-48-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR