8-Bromo-1-chloroisoquinoline

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Bromo-1-chloro-2-azanaphthalene

OR300069-100MG OR300069-250MG
APOSOR300069-100MGEA 21.9 EUR
APOSOR300069-100MG APOSOR300069-250MG
8-Bromo-1-chloroisoquinoline
8-Bromo-1-chloroisoquinoline
Formel: C₉H₅BrClN
Molecular Weight: 242,5 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13193666
CAS-Nummer: 1233025-78-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR