1-(Isoquinolin-3-yl)ethanone
Lieferant: Apollo Scientific
OR300098-250MG
OR300098-1G
APOSOR300098-250MGEA
94.4
EUR
APOSOR300098-250MG
APOSOR300098-1G
1-(Isoquinolin-3-yl)ethanone
1-(Isoquinolin-3-yl)ethanone
Formel:
C₁₁H₉NO Molecular Weight: 171.195 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06658291 CAS-Nummer: 91544-03-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...