Methyl 1-chloroisoquinoline-6-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Chloro-6-(methoxycarbonyl)isoquinoline, Methyl 1-chloro-2-azanaphthalene-6-carboxylate

OR300107-500MG OR300107-100MG
APOSOR300107-500MGEA 464 EUR
APOSOR300107-500MG APOSOR300107-100MG
Methyl 1-chloroisoquinoline-6-carboxylate 95%
Methyl 1-chloroisoquinoline-6-carboxylate
Formel: C₁₁H₈ClNO₂
Molecular Weight: 221,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21365100
CAS-Nummer: 1357946-43-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR