Methyl 5-bromoisoquinoline-1-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-1-(methoxycarbonyl)-2-azanaphthalene, 5-Bromo-1-(methoxycarbonyl)isoquinoline
OR300109-250MG
OR300109-100MG
APOSOR300109-250MGEA
638
EUR
APOSOR300109-250MG
APOSOR300109-100MG
Methyl 5-bromoisoquinoline-1-carboxylate
Methyl 5-bromoisoquinoline-1-carboxylate
Formel:
C₁₁H₈BrNO₂ Molecular Weight: 266,091 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19981746 CAS-Nummer: 1330750-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...