3-Chloro-6-hydroxyisoquinoline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloroisoquinolin-6-ol, 3-Chloro-6-hydroxy-2-azanaphthalene
OR300116-1G
OR300116-250MG
APOSOR300116-1GEA
899
EUR
APOSOR300116-1G
APOSOR300116-250MG
3-Chloro-6-hydroxyisoquinoline
3-Chloro-6-hydroxyisoquinoline
Formel:
C₉H₆ClNO Molecular Weight: 179,603 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22380493 CAS-Nummer: 1374652-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...