(Isoquinolin-5-yl)methanamine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR300123-500MG
OR300123-1G
APOSOR300123-500MGEA
97.3
EUR
APOSOR300123-500MG
APOSOR300123-1G
(Isoquinolin-5-yl)methanamine hydrochloride
(Isoquinolin-5-yl)methanamine hydrochloride
Formel:
C₁₀H₁₀N₂·ClH Molecular Weight: 194.661 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10696103 CAS-Nummer: 1001906-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...