1-Amino-6-(hydroxymethyl)isoquinoline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(1-Aminoisoquinolin-6-yl)methanol, 1-Amino-6-(hydroxymethyl)-2-azanaphthalene
OR300127-250MG
OR300127-100MG
APOSOR300127-250MGEA
435
EUR
APOSOR300127-250MG
APOSOR300127-100MG
1-Amino-6-(hydroxymethyl)isoquinoline
1-Amino-6-(hydroxymethyl)isoquinoline
Formel:
C₁₀H₁₀N₂O Molecular Weight: 174.199 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD22689644 CAS-Nummer: 1374656-02-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...