Methyl 3-chloroisoquinoline-6-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-6-(methoxycarbonyl)-2-azanaphthalene, 3-Chloro-6-(methoxycarbonyl)isoquinoline

OR300130-100MG OR300130-250MG
APOSOR300130-100MGEA 324 EUR
APOSOR300130-100MG APOSOR300130-250MG
Methyl 3-chloroisoquinoline-6-carboxylate 95%
Methyl 3-chloroisoquinoline-6-carboxylate
Formel: C₁₁H₈ClNO₂
Molecular Weight: 221,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22689680
CAS-Nummer: 1416713-88-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR