1-Iodo-7-methoxyisoquinoline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Iodo-7-methoxy-2-azanaphthalene
OR300132-500MG
OR300132-100MG
OR300132-250MG
APOSOR300132-500MGEA
277
EUR
APOSOR300132-500MG
APOSOR300132-100MG
APOSOR300132-250MG
1-Iodo-7-methoxyisoquinoline
1-Iodo-7-methoxyisoquinoline
Formel:
C₁₀H₈INO Molecular Weight: 285.081 g/mol Schmelzpunkt: 86…87 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04037478 CAS-Nummer: 952569-54-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...