3-Chloroisoquinoline-8-boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Borono-3-chloro-2-azanaphthalene, 8-Borono-3-chloroisoquinoline

OR300154-500MG OR300154-250MG OR300154-1G
APOSOR300154-500MGEA 498 EUR
APOSOR300154-500MG APOSOR300154-250MG APOSOR300154-1G
3-Chloroisoquinoline-8-boronic acid
3-Chloroisoquinoline-8-boronic acid
Formel: C₉H₇BClNO₂
Molecular Weight: 207,421 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22199344
CAS-Nummer: 2096333-80-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR