4-(3-Aminobicyclo[1.1.1]pentan-1-yl)phenol

Lieferant: Apollo Scientific
Achtung

OR300178-100MG OR300178-250MG
APOSOR300178-100MGEA 261 EUR
APOSOR300178-100MG APOSOR300178-250MG
4-(3-Aminobicyclo[1.1.1]pentan-1-yl)phenol
4-(3-Aminobicyclo[1.1.1]pentan-1-yl)phenol
Formel: C₁₁H₁₃NO
Molecular Weight: 175,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31536716
CAS-Nummer: 2209086-85-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR