1-Isothiocyanatobicyclo[1.1.1]pentane

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-isothiocyanatobicyclo[1.1.1]pentane

OR300179-250MG OR300179-1G
APOSOR300179-250MGEA 299 EUR
APOSOR300179-250MG APOSOR300179-1G
1-Isothiocyanatobicyclo[1.1.1]pentane
1-Isothiocyanatobicyclo[1.1.1]pentane
Formel: C₆H₇NS
Molecular Weight: 125,191 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD31536717
CAS-Nummer: 2187435-29-4
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR