3-Phenylpyrazin-2-amine
Lieferant: Apollo Scientific
|
Achtung
|
OR300239-1G
OR300239-250MG
APOSOR300239-1GEA
248
EUR
APOSOR300239-1G
APOSOR300239-250MG
3-Phenylpyrazin-2-amine
3-Phenylpyrazin-2-amine
Formel:
C₁₀H₉N₃ Lagertemperatur: Kühlschrank |
MDL:
MFCD05722803 CAS-Nummer: 41270-67-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...