5-Iodo-3-methylpyrazin-2-amine
Lieferant: Apollo Scientific
|
Achtung
|
OR300252-1G
OR300252-250MG
APOSOR300252-1GEA
488
EUR
APOSOR300252-1G
APOSOR300252-250MG
5-Iodo-3-methylpyrazin-2-amine
5-Iodo-3-methylpyrazin-2-amine
Formel:
C₅H₆IN₃ Molecular Weight: 235,026 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08705772 CAS-Nummer: 91416-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...