tert-Butyl 5-formylindoline-1-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Boc-5-formylindoline
OR300270-1G
OR300270-250MG
APOSOR300270-1GEA
290
EUR
APOSOR300270-1G
APOSOR300270-250MG
tert-Butyl 5-formylindoline-1-carboxylate
tert-Butyl 5-formylindoline-1-carboxylate
Formel:
C₁₄H₁₇NO₃ Molecular Weight: 247.29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12408236 CAS-Nummer: 879887-32-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...