tert-Butyl 2-bromo-6,7-dihydrothiazolo[4,5-c]pyridine-5(4H)-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

OR300276-1G OR300276-250MG OR300276-100MG
APOSOR300276-1GEA 353 EUR
APOSOR300276-1G APOSOR300276-250MG APOSOR300276-100MG
tert-Butyl 2-bromo-6,7-dihydrothiazolo[4,5-c]pyridine-5(4H)-carboxylate 95%
tert-Butyl 2-bromo-6,7-dihydrothiazolo[4,5-c]pyridine-5(4H)-carboxylate
Formel: C₁₁H₁₅BrN₂O₂S
Molecular Weight: 319,218 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17215781
CAS-Nummer: 1253654-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR