(5-Bromo-6-chloropyridin-3-yl)methanamine

Lieferant: Apollo Scientific
Gefahr

OR300318-100MG OR300318-250MG
APOSOR300318-100MGEA 174 EUR
APOSOR300318-100MG APOSOR300318-250MG
(5-Bromo-6-chloropyridin-3-yl)methanamine
(5-Bromo-6-chloropyridin-3-yl)methanamine
Formel: C₆H₆BrClN₂
Molecular Weight: 221,482 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18257860
CAS-Nummer: 1211592-59-4
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR