2,7-Dibromoquinoline
Lieferant: Apollo Scientific
|
Achtung
|
OR300365-1G
OR300365-250MG
APOSOR300365-1GEA
357
EUR
APOSOR300365-1G
APOSOR300365-250MG
2,7-Dibromoquinoline
2,7-Dibromoquinoline
Formel:
C₉H₅Br₂N Molecular Weight: 286,951 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09261125 CAS-Nummer: 1334405-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...