Methyl 5-bromoquinoline-8-carboxylate

Lieferant: Apollo Scientific
Achtung

OR300366-1G OR300366-250MG
APOSOR300366-1GEA 203 EUR
APOSOR300366-1G APOSOR300366-250MG
Methyl 5-bromoquinoline-8-carboxylate
Methyl 5-bromoquinoline-8-carboxylate
Formel: C₁₁H₈BrNO₂
Molecular Weight: 266,091 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19703294
CAS-Nummer: 1445781-45-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR