3-Bromo-5-chloro-1H-pyrazolo[3,4-b]pyridine
Lieferant: Apollo Scientific
|
Achtung
|
OR300379-1G
OR300379-250MG
APOSOR300379-1GEA
549
EUR
APOSOR300379-1G
APOSOR300379-250MG
3-Bromo-5-chloro-1H-pyrazolo[3,4-b]pyridine
3-Bromo-5-chloro-1H-pyrazolo[3,4-b]pyridine
Formel:
C₆H₃BrClN₃ Molecular Weight: 232,465 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14706800 CAS-Nummer: 1245649-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...