1-(2-Bromo-5-methylphenyl)ethan-1-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Bromo-5-methylphenyl)ethanone

OR300388-250MG OR300388-500MG
APOSOR300388-250MGEA 65.4 EUR
APOSOR300388-250MG APOSOR300388-500MG
1-(2-Bromo-5-methylphenyl)ethan-1-one
1-(2-Bromo-5-methylphenyl)ethan-1-one
Formel: C₉H₉BrO
Molecular Weight: 213.071 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11695376
CAS-Nummer: 77344-70-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR