3-Bromoquinoline-5-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR300389-1G OR300389-250MG
APOSOR300389-1GEA 377 EUR
APOSOR300389-1G APOSOR300389-250MG
3-Bromoquinoline-5-carboxylic acid
3-Bromoquinoline-5-carboxylic acid
Formel: C₁₀H₆BrNO₂
Molecular Weight: 252,064 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20327981
CAS-Nummer: 1344046-12-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR