2-Bromo-5-methyl-3-(methylthio)pyridine

Lieferant: Apollo Scientific
Achtung

OR300396-1G OR300396-250MG
APOSOR300396-1GEA 255 EUR
APOSOR300396-1G APOSOR300396-250MG
2-Bromo-5-methyl-3-(methylthio)pyridine
2-Bromo-5-methyl-3-(methylthio)pyridine
Formel: C₇H₈BrNS
Molecular Weight: 218,114 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26127742
CAS-Nummer: 1820649-46-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR