5-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazolo[3,4-b]pyridine

Lieferant: Apollo Scientific
Achtung

OR300398-1G OR300398-250MG
APOSOR300398-1GEA 345 EUR
APOSOR300398-1G APOSOR300398-250MG
5-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazolo[3,4-b]pyridine
5-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazolo[3,4-b]pyridine
Formel: C₁₁H₁₂BrN₃O
Molecular Weight: 282,136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22689759
CAS-Nummer: 1256957-72-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR