Ethyl 2,3-dioxopiperidine-4-carboxylate
Lieferant: Apollo Scientific
Synonyme:
Ethyl 5-hydroxy-6-oxo-1,2,3,6-tetrahydropyridine-4-carboxylate
OR300500-1G
OR300500-250MG
OR300500-500MG
APOSOR300500-1GEA
510
EUR
APOSOR300500-1G
APOSOR300500-250MG
APOSOR300500-500MG
Ethyl 2,3-dioxopiperidine-4-carboxylate
Ethyl 2,3-dioxopiperidine-4-carboxylate
Formel:
C₈H₁₁NO₄ Molecular Weight: 185.177 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00185013 CAS-Nummer: 30727-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...