Ethyl 5-phenyl-1,3,4-oxadiazole-2-carboxylate
Lieferant: Apollo Scientific
Synonyme:
5-Phenyl-1,3,4-oxadiazole-2-carboxylic acid ethyl ester
OR300512-1G
OR300512-2.5G
OR300512-5G
OR300512-10G
APOSOR300512-1GEA
191
EUR
APOSOR300512-1G
APOSOR300512-2.5G
APOSOR300512-5G
APOSOR300512-10G
Ethyl 5-phenyl-1,3,4-oxadiazole-2-carboxylate
Ethyl 5-phenyl-1,3,4-oxadiazole-2-carboxylate
Formel:
C₁₁H₁₀N₂O₃ Molecular Weight: 218,209 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03753690 CAS-Nummer: 16691-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...