3-(2-Methylthiazol-4-yl)nitrobenzene
Lieferant: Apollo Scientific
Synonyme:
2-Methyl-4-(3-nitrophenyl)thiazole
OR300557-1G
OR300557-250MG
OR300557-500MG
OR300557-2.5G
APOSOR300557-1GEA
308
EUR
APOSOR300557-1G
APOSOR300557-250MG
APOSOR300557-500MG
APOSOR300557-2.5G
3-(2-Methylthiazol-4-yl)nitrobenzene
3-(2-Methylthiazol-4-yl)nitrobenzene
Formel:
C₁₀H₈N₂O₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00838954 CAS-Nummer: 39541-91-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...