Ethyl 3-methyl-5-phenylpyrazole-4-carboxylate
Lieferant: Apollo Scientific
Synonyme:
Ethyl 5-methyl-3-phenyl-1H-pyrazole-4-carboxylate
OR300580-250MG
OR300580-500MG
APOSOR300580-250MGEA
171
EUR
APOSOR300580-250MG
APOSOR300580-500MG
Ethyl 3-methyl-5-phenylpyrazole-4-carboxylate
Ethyl 3-methyl-5-phenylpyrazole-4-carboxylate
Formel:
C₁₃H₁₄N₂O₂ Molecular Weight: 230,262 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659797 CAS-Nummer: 76923-16-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...