2,3,5-Trichlorophenylacetonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2,3,5-Trichlorophenyl)acetonitrile, 2,3,5-Trichlorobenzyl cyanide
OR300722-250MG
OR300722-500MG
APOSOR300722-250MGEA
223
EUR
APOSOR300722-250MG
APOSOR300722-500MG
2,3,5-Trichlorophenylacetonitrile
2,3,5-Trichlorophenylacetonitrile
Formel:
C₈H₄Cl₃N Molecular Weight: 220,483 g/mol Schmelzpunkt: 182…184 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09923728 CAS-Nummer: 4681-16-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...