Ethyl 3-(4-benzylpiperidin-1-yl)propionate
Lieferant: Apollo Scientific
OR300893-1G
OR300893-5G
APOSOR300893-1GEA
0
EUR
APOSOR300893-1G
APOSOR300893-5G
Ethyl 3-(4-benzylpiperidin-1-yl)propionate
Ethyl 3-(4-benzylpiperidin-1-yl)propionate
Formel:
C₁₇H₂₅NO₂ Molecular Weight: 275.386 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13633484 CAS-Nummer: 1099610-66-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...