1,1-Diethylguanidine hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR300900-1G OR300900-250MG
APOSOR300900-1GEA 0 EUR
APOSOR300900-1G APOSOR300900-250MG
1,1-Diethylguanidine hydrochloride 95%
1,1-Diethylguanidine hydrochloride
Formel: C₅H₁₃N₃·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD00035042
CAS-Nummer: 1114-39-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR