1-Phenethylguanidine hydrochloride

Lieferant: Apollo Scientific

OR300902-1G OR300902-250MG
APOSOR300902-1GEA 0 EUR
APOSOR300902-1G APOSOR300902-250MG
1-Phenethylguanidine hydrochloride
1-Phenethylguanidine hydrochloride
Formel: C₉H₁₃N₃·ClH
Molecular Weight: 199,681 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09759100
CAS-Nummer: 2235-99-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR