6-Bromo-1H-indazole-3-carbaldehyde 97%

Lieferant: Apollo Scientific
Achtung

OR301105-1G OR301105-5G OR301105-250MG
APOSOR301105-1GEA 45.1 EUR
APOSOR301105-1G APOSOR301105-5G APOSOR301105-250MG
6-Bromo-1H-indazole-3-carbaldehyde 97%
6-Bromo-1H-indazole-3-carbaldehyde
Formel: C₈H₅BrN₂O
Molecular Weight: 225,042 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06738286
CAS-Nummer: 885271-72-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR